Autocamtide-2-Related Inhibitory Peptide
Catalog # H-3384.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:167114-91-2
- Source:
- Species:
- Size:1 mg
- Environmentally Preferable:
- Storage Conditions:–20 ± 5 °C
- Protein Synonyms:AIP|(Ala9)-Autocamtide-2
- Protein/Peptide Name:Autocamtide-2-Related Inhibitory Peptide
- Molecular Weight:1497.76
- Sequence:H-Lys-Lys-Ala-Leu-Arg-Arg-Gln-Glu-Ala-Val-Asp-Ala-Leu-OH
- Molecular Formula:C₆₄H₁₁₆N₂₂O₁₉
- Cat. No.:H-3384.0001BA
- Supplier no.:H-3384.0001
Specifications
About this item
AIP, non-phosphorylatable analog of autocamtide-2 (H-3218), was found to be a highly specific and potent inhibitor of calmodulin-dependent protein Kinase II (IC₅₀= 40 nM) without affecting cyclic AMP-dependent protein kinase, protein kinase C and calmodulin-dependent protein kinase IV.