Order Entry
United States
ContactUsLinkComponent
Human;Rat (D-Phe¹²,Nle²¹·³⁸,alpha-Me-Leu³⁷)-CRF (12-41) Trifluoroacetate
Human;Rat (D-Phe¹²,Nle²¹·³⁸,alpha-Me-Leu³⁷)-CRF (12-41) Trifluoroacetate
Supplier:  Bachem Americas
Certificates
Human;Rat (D-Phe¹²,Nle²¹·³⁸,alpha-Me-Leu³⁷)-CRF (12-41) Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    150646-45-0
  • Species:
    Human
    Rat
  • Storage Conditions:
    –20 ± 5 °C
  • Protein/Peptide Name:
    (D-Phe¹²,Nle²¹·³⁸,alpha-Me-Leu³⁷)-CRF (12-41) Trifluoroacetate
  • Molecular Weight:
    3553.17
  • Sequence:
    H-D-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Nle-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-Gln-Ala-His-Ser-Asn-Arg-Lys-α-Me-Leu-Nle-Glu-Ile-Ile-NH₂
  • Molecular Formula:
    C₁₅₉H₂₆₇N₄₉O₄₃

Specifications

Frequently Bought Together