H-Glu-Glu-Glu-OH
Supplier: Bachem Americas
Certificates
About this item
Triglutamic acid, H-3160, was shown to increase the release of vasodilatory substances as PGI₂ and NO.
Specifications
- CAS No.:23684-48-2
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00037310
- Protein/Peptide Name:H-Glu-Glu-Glu-OH
- Molecular Weight:405.36
- Sequence:H-Glu-Glu-Glu-OH
- Molecular Formula:C₁₅H₂₃N₃O₁₀
Specifications
Frequently Bought Together