Human Recombinant Growth Hormone (from E. coli)
Supplier: Bachem Americas
Certificates
About this item
Fully active in biological assays when compared to known standards. Endotoxin level < 0.1 ng per µg hGH.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:−20 ± 5 °C
- MDL No.:MFCD00677957
- Protein Synonyms:hGH
- Protein/Peptide Name:Growth Hormone
- Sequence:0
Specifications
Frequently Bought Together