HIV-1 gag Protein p24 (194-210)
H-2986.1000BA
:
- CAS No.:141281-67-6
- Source:
- Species:
- Size:1 mg
- Environmentally Preferable:
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00076829
- Protein/Peptide Name:HIV-1 gag Protein p24 (194-210)
- Molecular Weight:1683.99
- Sequence:H-Ala-Asn-Pro-Asp-Cys-Lys-Thr-Ile-Leu-Lys-Ala-Leu-Gly-Pro-Ala-Ala-Thr-OH
- Molecular Formula:C₇₃H₁₂₆N₂₀O₂₃S
- Cat. No.:H-2986.1000BA
- Supplier no.:H-2986.1000