Leu-Enkephalin
Supplier: Bachem Americas
Certificates
About this item
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.
The sequence of Leu-enkephalin, YGGFL, corresponds to the N-terninus of dynorphin A.
N-terminal sequence of dynorphin A and the neoendorphins.
Specifications
- CAS No.:58822-25-6
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00076406
- Protein Synonyms:Dynorphin A (1-5)|a-Neoendorphin (1-5)
- Protein/Peptide Name:Leu-Enkephalin
- Molecular Weight:555.63
- Sequence:H-Tyr-Gly-Gly-Phe-Leu-OH
- Molecular Formula:C₂₈H₃₇N₅O₇
Specifications
Frequently Bought Together