Human alpha-Endorphin
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Dynorphin, Analogs and Sequences, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides’.
- CAS No.:59004-96-5
- Species:Human
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00076381
- Protein Synonyms:b-Endorphin (1-16)|b-Lipotropin (59-74)
- Protein/Peptide Name:alpha-Endorphin
- Molecular Weight:1745.97
- Sequence:H-Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-OH
- Molecular Formula:C₇₇H₁₂₀N₁₈O₂₆S
Frequently Bought Together