Order Entry
United States
ContactUsLinkComponent
IGF-I (1-3)
IGF-I (1-3)
Supplier:  Bachem Americas
Certificates
IGF-I (1-3)
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    32302-76-4
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00144405
  • Protein Synonyms:
    GPE
  • Protein/Peptide Name:
    IGF-I (1-3)
  • Molecular Weight:
    301.30
  • Sequence:
    H-Gly-Pro-Glu-OH
  • Molecular Formula:
    C₁₂H₁₉N₃O₆

Specifications

Frequently Bought Together