Order Entry
United States
ContactUsLinkComponent
Stichodactyla Helianthus Neurotoxin (ShK) Trifluoroacetate
Stichodactyla Helianthus Neurotoxin (ShK) Trifluoroacetate
Supplier:  Bachem Americas
Certificates
Stichodactyla Helianthus Neurotoxin (ShK) Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    172450-46-3
  • Storage Conditions:
    –20 ± 5 °C
  • Protein Synonyms:
    ShK
  • Protein/Peptide Name:
    Stichodactyla helianthus Neurotoxin (ShK) Trifluoroacetate
  • Molecular Weight:
    4054.83
  • Sequence:
    H-Arg-Ser-Cys-Ile-Asp-Thr-Ile-Pro-Lys-Ser-Arg-Cys-Thr-Ala-Phe-Gln-Cys-Lys-His-Ser-Met-Lys-Tyr-Arg-Leu-Ser-Phe-Cys-Arg-Lys-Thr-Cys-Gly-Thr-Cys-OH
  • Molecular Formula:
    C₁₆₉H₂₇₄N₅₄O₄₈S₇

Specifications

Frequently Bought Together