Order Entry
United States
ContactUsLinkComponent
Human;Mouse;Rat (Phe¹³,Tyr¹⁹)-MCH Trifluoroacetate
Human;Mouse;Rat (Phe¹³,Tyr¹⁹)-MCH Trifluoroacetate
Supplier:  Bachem Americas
Certificates
Human;Mouse;Rat (Phe¹³,Tyr¹⁹)-MCH Trifluoroacetate
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    160201-86-5
  • Species:
    Human
    Rat
    Mouse
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD18782134
  • Protein Synonyms:
    (Phe13|Tyr19)-Melanin-Concentrating Hormone (MCH)
  • Protein/Peptide Name:
    (Phe¹³,Tyr¹⁹)-MCH Trifluoroacetate
  • Molecular Weight:
    2434.92
  • Sequence:
    H-Asp-Phe-Asp-Met-Leu-Arg-Cys-Met-Leu-Gly-Arg-Val-Phe-Arg-Pro-Cys-Trp-Gln-Tyr-OH
  • Molecular Formula:
    C₁₀₉H₁₆₀N₃₀O₂₆S₄

Specifications

Frequently Bought Together