H-Arg-Arg-Arg-OH Acetate, Bachem
Supplier: Bachem Americas
Certificates
About this item
Triarginine.
Specifications
- CAS No.:6418-87-7
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD03788118
- Protein/Peptide Name:H-Arg-Arg-Arg-OH Acetate
- Molecular Weight:486.58
- Sequence:H-Arg-Arg-Arg-OH
- Molecular Formula:C₁₈H₃₈N₁₂O₄
Specifications
Frequently Bought Together