Order Entry
United States
ContactUsLinkComponent
H-Tyr-Trp-OH
H-Tyr-Trp-OH
  G-3470.0250BA
 :  
H-Tyr-Trp-OH
  G-3470.0250BA
 :  4000897.0250
 :  

 

  • CAS No.:
    60815-41-0
  • Size:
    250 mg
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00083832
  • Protein/Peptide Name:
    H-Tyr-Trp-OH
  • Molecular Weight:
    367.41
  • Sequence:
    H-Tyr-Trp-OH
  • Molecular Formula:
    C₂₀H₂₁N₃O₄
  • Cat. No.:
    G-3470.0250BA
  • Supplier no.:
    G-3470.0250