Order Entry
United States
Orders LinkContactUsLinkComponent
H-Pro-Met-OH

H-Pro-Met-OH

1 g
Catalog # G-3045.0001BA
Supplier:   Bachem Americas
Supplier #:  4014003.0001
H-Pro-Met-OH
Catalog # G-3045.0001BA
Supplier:  Bachem Americas
CAS Number:  
H-Pro-Met-OH

1 g

CAS Number:  
Price
$574.98
Log in for availability
Each (1g)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Substrate for prolinase (prolyl dipeptidase).

Specifications

Specifications

  • Size:
    1 g
  • Storage Conditions:
    –20 ± 5 °C
  • Cat. No.:
    G-3045.0001BA
  • Molecular Weight:
    246.33
  • Protein/Peptide Name:
    H-Pro-Met-OH
  • MDL No.:
    MFCD00057180
  • Sequence:
    H-Pro-Met-OH
  • Molecular Formula:
    C₁₀H₁₈N₂O₃S
  • CAS No.:
    52899-08-8

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.