Order Entry
United States
ContactUsLinkComponent
H-Phe-Trp-OH
H-Phe-Trp-OH
Catalog # G-2950.0001BA
Supplier:  Bachem Americas
CAS Number:  
H-Phe-Trp-OH
Catalog # G-2950.0001BA
Supplier:  Bachem Americas
Supplier Number:  4005237.0001
CAS Number:  

Specifications

  • CAS No.:
    24587-41-5
  • Size:
    1 g
  • Storage Conditions:
    –20 ± 5 °C
  • Protein/Peptide Name:
    H-Phe-Trp-OH
  • Molecular Weight:
    351.41
  • Sequence:
    H-Phe-Trp-OH
  • Molecular Formula:
    C₂₀H₂₁N₃O₃
  • Cat. No.:
    G-2950.0001BA
  • Supplier no.:
    G-2950.0001

Specifications

About this item

These short peptides act as angiotensin I-converting enzyme (ACE) inhibitors. See also Phe-Tyr (G-2955) and the subfamilies 'Bradykinin-Potentiating Peptide (BPP)' and 'Angiotensin I-Converting Enzyme (ACE, ACE2) Inhibitors'.

FW, a highly potent ACE inhibitor, can also form nanotubes.