Order Entry
United States
ContactUsLinkComponent
H-Gly-Gly-Phe-OH
H-Gly-Gly-Phe-OH
Catalog # H-3465.0005BA
Supplier:  Bachem Americas
CAS Number:  
H-Gly-Gly-Phe-OH
Catalog # H-3465.0005BA
Supplier:  Bachem Americas
Supplier Number:  4002475.0005
CAS Number:  

Specifications

  • CAS No.:
    6234-26-0
  • Size:
    5 g
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD00066054
  • Protein/Peptide Name:
    H-Gly-Gly-Phe-OH
  • Molecular Weight:
    279.30
  • Sequence:
    H-Gly-Gly-Phe-OH
  • Molecular Formula:
    C₁₃H₁₇N₃O₄
  • Cat. No.:
    H-3465.0005BA
  • Supplier no.:
    H-3465.0005

Specifications

About this item

For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.

Enkephalin (2-4), GGF, stimulated the uptake of deltorphin in SK-N-SH cells.