Order Entry
United States
ContactUsLinkComponent
Carcinine
Carcinine
Supplier:  Bachem Americas
Certificates
Carcinine
Supplier:  Bachem Americas

Specifications

  • CAS No.:
    56897-53-1
  • Storage Conditions:
    –20 ± 5 °C
  • MDL No.:
    MFCD01310858
  • Protein/Peptide Name:
    Carcinine
  • Molecular Weight:
    182.23
  • Sequence:
    H-β-Ala-histamine
  • Molecular Formula:
    C₈H₁₄N₄O

Specifications

Frequently Bought Together