H-Lys-Gly-OH · HCl
Supplier: Bachem Americas
Certificates
About this item
Specifications
- CAS No.:40719-58-2
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00039066
- Protein/Peptide Name:H-Lys-Gly-OH · HCl
- Molecular Weight:239.70
- Sequence:H-Lys-Gly-OH · HCl
- Molecular Formula:C₈H₁₇N₃O₃ · HCl
Specifications
Frequently Bought Together