H-Gly-D-Phe-OH
Catalog # G-2180.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- CAS No.:34258-14-5
- Size:5 g
- Storage Conditions:–20 ± 5 °C
- MDL No.:MFCD00025584
- Protein/Peptide Name:H-Gly-D-Phe-OH
- Molecular Weight:222.24
- Sequence:H-Gly-D-Phe-OH
- Molecular Formula:C₁₁H₁₄N₂O₃
- Cat. No.:G-2180.0005BA
- Supplier no.:G-2180.0005
Specifications
About this item
Substrate for D-peptidase S, an intracellular carboxypeptidase-like enzyme that preferentially hydrolyzes dipeptides containing hydrophobic D-amino acids.