Orders LinkContactUsLinkComponent
Universal Clamp Holder, Jumbo
Jumbo Universal Clamp Holder, 20 mm
  470137-836
 :   Wards
 :  CH0664JM
Universal Clamp Holder, Jumbo
  470137-836
 :  Wards
Universal Clamp Holder, Jumbo
Universal Clamp Holder, Jumbo
Jumbo Universal Clamp Holder, 20 mm
$11.95
Each
Large Clamp Capacity You Need.
  • Accommodates rods and flat boards
This larger clamp holder accommodates rods up to 20mm in diameter and flat boards up to 18mm thick.
  • Description:
    Jumbo Universal Clamp Holder
  • Rod diameter:
    20 mm