ContactUsLinkComponent
Eisco® Boss Head, Aluminum
Eisco® Boss Head, Aluminum
Catalog # 470137-834
Supplier:  Wards
undefined
Eisco® Boss Head, Aluminum
Catalog # 470137-834
Supplier:  Wards
Supplier Number:  CH0664A
Specifications
  • Description:
    Boss Head - Aluminum
  • Environmentally Preferable:
  • Cat. No.:
    470137-834
  • Supplier no.:
    CH0664A
Specifications
About this item
Secure laboratory equipment and apparatus at right angles.
  • Heavy duty, aluminum die cast
  • Accepts rods up to 16 mm (Ø)
  • Holds rods at right angles
This die cast aluminum boss head holds rods securely at right angles. The metal thumbscrews are durable, easy-to-use and provide a very secure grip.