ContactUsLinkComponent
Solderless Breadboard
Catalog # 470137-790
undefined
Solderless Breadboard
Catalog # 470137-790
Supplier Number:  GS-830T
Specifications
  • Description:
    Transparent Breadboard, External Power 830 Tie Points
  • Cat. No.:
    470137-790
  • Supplier no.:
    GS-830T
Specifications
About this item
Low Cost Meets Quality.
  • 830 tie points
  • Size 6.5×2.1"
This solderless breadboarding system has 830 tie points, and allows students to perform lab experiments in all areas of custom electronic design applications and to design test fixtures. The color coded rails and clearly labeled grid allow for easy viewing and help students understand the conductive pathways.