Order Entry
United States
ContactUsLinkComponent
Human alpha-Endorphin
Human alpha-Endorphin
Catalog # AAJ63462-MCR
CAS Number:  
Human alpha-Endorphin
Catalog # AAJ63462-MCR
Supplier Number:  J63462-MCR
CAS Number:  

Specifications

  • CAS No.:
    59004-96-5
  • Species:
    Human
  • Size:
    1 mg
  • Storage Conditions:
    Ambient
  • Form:
    Powder
  • MDL No.:
    MFCD00076381
  • Protein/Peptide Name:
    alpha-Endorphin
  • Molecular Weight:
    1745.98
  • Color:
    White
  • Molecular Formula:
    C₇₇H₁₂₀N₁₈O₂₆S
  • Cat. No.:
    AAJ63462-MCR
  • Supplier no.:
    J63462-MCR

Specifications

About this item

CAS No.: 59004-96-5
Molecular Formula: C77H120N18O26S
Formula Weight: 1745.98
Physical Form: Powder
Appearance: White

An opioid peptide and substrate for transglutaminase