LB Agar Plate with Kanamycin, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
This LB Agar plate with kanamycin is suitable for selective cultivation of E.coli strains that contain plasmids conferring kanamycin resistance.
- Solid
- Not miscible or difficult to mix in water
The bacteria can be used for DNA plasmid production or production of recombinant proteins.
Caution: Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications
- Storage Conditions:2 - 8 °C