ContactUsLinkComponent
Flask, Two Neck, Pear-Shaped, Microscale, Ace Glass Incorporated
Flask, Two Neck, Pear-Shaped, Microscale, Ace Glass Incorporated
Catalog # 89074-922
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Flask, Two Neck, Pear-Shaped, Microscale, Ace Glass Incorporated
Catalog # 89074-922
Supplier:  Ace Glass
Supplier Number:  9479-29
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Capacity:
    25 ml
  • Cat. No.:
    89074-922
  • Supplier no.:
    9479-29
Specifications
About this item
These microscale, pear-shaped flasks have joints that are a combination cap, thread and [ST] joint to accommodate and seal other microscale apparatus
The O.D. of the joints have an external thread to facilitate an o-ring and an open-top phenolic cap. The internal (or I.D.) at the top is a ground [ST] 14/10 outer joint. The flask is supplied with a holed cap, PTFE-faced septa, and FETFE o-rings. Fabricated from borosilicate glass.