Order Entry
United States
ContactUsLinkComponent
Corning® Poloxamer, Corning
Corning® Poloxamer, Corning
Supplier:  Corning
Certificates
undefined
Corning® Poloxamer, Corning
Supplier:  Corning
Certificates

Specifications

  • Shipping Conditions:
    Ambient
  • Storage Temperature:
    Refrigerate

Specifications

Related Information

Frequently Bought Together