Order Entry
United States
Orders LinkContactUsLinkComponent
Corning® MOPS Buffer, Powder, Corning

Corning® MOPS Buffer, Powder, Corning

MOPS Buffer, Powder, 100 g
Catalog # 45001-134
Supplier:   Corning
Supplier #:  46-103-RM
Corning® MOPS Buffer, Powder, Corning
Catalog # 45001-134
Supplier:  Corning
CAS Number:  
Corning® MOPS Buffer, Powder, Corning
Corning® MOPS Buffer, Powder, Corning

MOPS Buffer, Powder, 100 g

CAS Number:  
Price
$296.00
Log in for availability
Each (100g)
Minimum order quantity is 1
Product Details & Documents

Documents

Specifications

Specifications

  • Description:
    MOPS Buffer, Powder
  • Size:
    100 g
  • Cat. No.:
    45001-134
  • Shipping Temperature:
    Ambient
  • Storage Temperature:
    Ambient

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.