Order Entry
ContactUsLinkComponent
Corning® insectagro® DS2 Serum-Free/Protein-Free Medium, 1X without L-glutamine, Corning
Corning® insectagro® DS2 Serum-Free/Protein-Free Medium, 1X without L-glutamine, Corning
Catalog # 45000-474
Supplier:  Corning
CAS Number:  
undefined
Corning® insectagro® DS2 Serum-Free/Protein-Free Medium, 1X without L-glutamine, Corning
Catalog # 45000-474
Supplier:  Corning
Supplier Number:  13-402-CV
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Description:
    Corning® insectagro® DS2 Serum-Free/Protein-Free Medium, 1X without L-glutamine
  • Environmentally Preferable:
  • Storage Temperature:
    Refrigerate
  • Cat. No.:
    45000-474
  • Supplier no.:
    13-402-CV

Specifications

About this item

Corning® insectagro® DS2 Serum-Free/Protein-Free Medium was developed for the growth and maintenance of Drosophila Schneider 2 (S2) cells to be used in heterologous protein expression

At the optimal temperature range 22-24ºC, DS2 cells grow as a loose monolayer and are readily adaptable to growth in suspension. Under these conditions, the cells require minimal adaptation to serum-free culture.

Frequently Bought Together