Hygromycin B solution 50 mg/ml, sterile
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Hygromycin B, produced by Streptomyces hygroscopicus, is used as a selection agent and inhibits protein synthesis in cells not carrying hygromycin phosphotransferase (HPH)
Antibiotic; For selection/maintenance of prokaryotic, eukaryotic cells.
Application: Recommended for use as a selection agent at 100-800 μg/mL. Biochem/physiol Actions: Mode of Action: Blocks polypeptide synthesis and inhibits elongation. For use in the selection and maintenance of prokaryotic and eukaryotic cells. Preparation Note: Purified by ion exchange chromatography.
HPH inactivates hygromycin B and restores protein synthesis.
: Shipping temperature: Ambient
- CAS Number:31282-04-9
- Formula:C₂₀H₃₇N₃O₁₃
- MDL Number:MFCD06795479
- Molecular Weight:527.5 g/mol
- Storage Temperature:Refrigerator
Documents
Certificates
Search by Lot NumberCell Culture Media, Molecular Buffers and Reagents
Additional Product Details
Corning Cell Culture Media.pdf
Promotion Material
Specifications
Specifications
- Size:20 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



