Corning® Ham's F-10 Medium, Corning
Catalog # 45000-356
Supplier: Corning
CAS Number:
Specifications
- Media Format:Liquid
- Description:Ham's F-10 Medium with L-glutamine
- Storage Temperature:Refrigerate
- Cat. No.:45000-356
- Supplier no.:10-070-CV
Specifications
About this item
Ham's F-10 is a classical media designed to support the growth of a variety of cell types, including HeLa, Chinese Hamster Ovary (CHO), primary cells from tissue explants and various types of human diploid cells.
- Low endotoxin - < 0.25EU/mL
- Mycoplasma tested
- Formulated with cell culture grade WFI water
- Manufactured in a cGMP environment