High Resolution Micro-Ohm Meter, Extech
Supplier: Avantor
Certificates
About this item
Precision meter provides resolution down to 1µOhm with 6 ranges with 3 sub-ranges in each current range
Measures resistive and inductive materials. Meter has a 5-Digit backlit LCD, PC interface and can store/recall up to 3000 measurements.
Certifications: CE marked.
Ordering information: Comes with 4-wire cables with Kelvin clip connectors, rechargeable Li-Polymer battery, AC adapter/charger, software and USB cable, and soft carrying case. 1-Year Warranty.
Frequently Bought Together