Order Entry
United States
ContactUsLinkComponent
Utility Clamp, Ace Glass
Utility Clamp, Ace Glass
Catalog # 89083-218
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Utility Clamp, Ace Glass
Catalog # 89083-218
Supplier:  Ace Glass
Supplier Number:  11060-13
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Description:
    Utility Clamp, Vinylized Jaws
  • Cat. No.:
    89083-218
  • Supplier no.:
    11060-13

Specifications

About this item

This utility clamp with vinylized jaws can be extended in length 13mm and firmly locked in any position

The unique safety-lock wing nut can be securely tightened, and the jaws will not swivel under strain. This clamp will hold objects up to 38mm in diameter, and will grip rods up to 19mm in diameter.

Ordering information: Comes complete with an extra set of fiberglass prong covers for temperatures above 100°C.