Orders LinkContactUsLinkComponent
MEGA FlexTubes, IBI Scientific
Supplier:   IBI Scientific
MEGA FlexTubes, IBI Scientific
Supplier:  IBI Scientific
Certificates
MEGA FlexTubes, IBI Scientific
MEGA FlexTubes, IBI Scientific
37 Options Available Below
Product Details & Documents
IBI MEGA FlexTubes are typically used for dialysis or buffer exchange with volumes of 3 to 20 ml.
The FlexTube membrane is ultra-clean, sulfur- and heavy metal-free, and EDTA treated.
IBI FlexTubes allow rapid, secure, and simple loading and recovery of samples. IBI FlexTubes can also be used for applications such as the preparation of protein samples for MALDI-MS, sample concentration, tissue culture extraction purification, removal of salts, surfactants, solvents, and detergents, as well as peptide dialysis and virus-particle purification. The performance of IBI MEGA FlexTubes are regularly monitored and tested by simple dialysis and assays commonly used for proteins and/or nucleic acids.
Product Family Options
Product Information
  • Volume
    MWCO
    Availability
    Price
  • 89369-600
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    10 ml
    1 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $198.52
    $198.52
    Pack of 5
       $198.52
    Supplier Number: IB48310
  • 89369-602
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    10 ml
    1 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $558.79
    $558.79
    Pack of 15
       $558.79
    Supplier Number: IB48311
  • 89369-604
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    15 ml
    1 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $198.52
    $198.52
    Pack of 5
       $198.52
    Supplier Number: IB48320
  • 89369-606
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    15 ml
    1 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $558.79
    $558.79
    Pack of 15
       $558.79
    Supplier Number: IB48321
  • 89369-608
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    20 ml
    1 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $198.52
    $198.52
    Pack of 5
       $198.52
    Supplier Number: IB48330
  • 89369-610
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    20 ml
    1 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $558.79
    $558.79
    Pack of 15
       $558.79
    Supplier Number: IB48331
  • 89369-594
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    10 ml
    1 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $77.90
    $77.90
    Pack of 2
       $77.90
    Supplier Number: IB48300
  • 89369-596
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    15 ml
    1 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $77.90
    $77.90
    Pack of 2
       $77.90
    Supplier Number: IB48301
  • 89369-598
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    20 ml
    1 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $77.90
    $77.90
    Pack of 2
       $77.90
    Supplier Number: IB48302
  • 89369-618
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    10 ml
    3.5 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $98.57
    $98.57
    Pack of 5
       $98.57
    Supplier Number: IB48350
  • 89369-620
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    10 ml
    3.5 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $257.34
    $257.34
    Pack of 15
       $257.34
    Supplier Number: IB48351
  • 89369-622
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    15 ml
    3.5 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $98.57
    $98.57
    Pack of 5
       $98.57
    Supplier Number: IB48360
  • 89369-624
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    15 ml
    3.5 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $257.34
    $257.34
    Pack of 15
       $257.34
    Supplier Number: IB48361
  • 89369-626
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    20 ml
    3.5 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $98.57
    $98.57
    Pack of 5
       $98.57
    Supplier Number: IB48370
  • 89369-628
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    20 ml
    3.5 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $259.60
    $259.60
    Pack of 15
       $259.60
    Supplier Number: IB48371
  • 89369-612
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    10 ml
    3.5 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48340
  • 89369-614
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    15 ml
    3.5 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48341
  • 89369-616
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    20 ml
    3.5 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48342
  • 89369-636
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    10 ml
    6 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $95.60
    $95.60
    Pack of 5
       $95.60
    Supplier Number: IB48390
  • 89369-638
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    10 ml
    6 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $257.34
    $257.34
    Pack of 15
       $257.34
    Supplier Number: IB48391
  • 89369-640
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    15 ml
    6 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $95.60
    $95.60
    Pack of 5
       $95.60
    Supplier Number: IB48400
  • 89369-642
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    15 ml
    6 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $257.34
    $257.34
    Pack of 15
       $257.34
    Supplier Number: IB48401
  • 89369-644
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    20 ml
    6 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $95.60
    $95.60
    Pack of 5
       $95.60
    Supplier Number: IB48410
  • 89369-646
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    20 ml
    6 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $257.34
    $257.34
    Pack of 15
       $257.34
    Supplier Number: IB48411
  • 89369-630
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    10 ml
    6 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48380
  • 89369-632
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    15 ml
    6 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48381
  • 89369-634
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    20 ml
    6 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48382
  • 89369-654
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    10 ml
    12 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $96.44
    $96.44
    Pack of 5
       $96.44
    Supplier Number: IB48430
  • 89369-656
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    10 ml
    12 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $257.34
    $257.34
    Pack of 15
       $257.34
    Supplier Number: IB48431
  • 89369-658
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    15 ml
    12 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $95.60
    $95.60
    Pack of 5
       $95.60
    Supplier Number: IB48440
  • 89369-660
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    15 ml
    12 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $257.34
    $257.34
    Pack of 15
       $257.34
    Supplier Number: IB48441
  • 89369-662
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    20 ml
    12 K
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $95.60
    $95.60
    Pack of 5
       $95.60
    Supplier Number: IB48450
  • 89369-664
    MEGA FlexTube Kit
    MEGA FlexTube Kit
    20 ml
    12 K
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $257.34
    $257.34
    Pack of 15
       $257.34
    Supplier Number: IB48451
  • 89369-648
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    10 ml
    12 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48420
  • 89369-650
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    15 ml
    12 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48421
  • 89369-652
    MEGA FlexTube Kit, Float
    MEGA FlexTube Kit, Float
    20 ml
    12 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48422
  • 89369-456
    MEGA FlexTube Float - 1 Place
    MEGA FlexTube Float - 1 Place
    Log in for availability
    Log in for availability
    Each
    Each - $26.44
    $26.44
    Each
       $26.44
    Supplier Number: IB48007