Orders LinkContactUsLinkComponent
MINI Flextubes, IBI Scientific
Supplier:   IBI Scientific
MINI Flextubes, IBI Scientific
Supplier:  IBI Scientific
Certificates
MINI Flextubes, IBI Scientific
MINI Flextubes, IBI Scientific
20 Options Available Below
Product Details & Documents
IBI MINI FlexTubes are typically used for the extraction of proteins, RNA, DNA, or oligonucleotides (>20 nt) from polyacrylamide, agarose, or any other gel matrix in a running buffer.
MINI FlexTubes require a minimum of 0.5 µg of protein at the start of extraction. The maximum size of the gel slice that can be inserted into the MINI FlexTube is 0.4×1.1 cm.
The yield of DNA/RNA recovery from an agarose gel or polyacrylamide gel is 90%. The yield of protein recovery from an SDS-PAGE gel is 70%. The FlexTube membrane is ultra-clean, sulfur- and heavy metal-free, and EDTA treated.
They are also used for dialysis or buffer exchange of small volumes (10 to 250 µl), preparation of protein samples for MALDI-MS, 2D gel dots extraction, and sample concentrations. IBI MINI FlexTubes are tested by using them in the extraction of proteins, RNA, or DNA fragments of various sizes from either agarose or polyacrylamide gels. IBI MINI FlexTube kits are also tested for simple dialysis of salts or buffer exchanges. The quality of the isolated protein, RNA, or DNA fragments, or of the sample after dialysis, is checked by several assays commonly used for proteins, nucleic acids, and dialysis. These devices combine two modes of action, electro-elution of macromolecules from polyacrylamide or agarose gels and buffer exchange of small volume samples. These devices allow rapid run times and high performance in either mode, and extract the macromolecules without contamination.
Product Family Options
Product Information
  • Volume
    For sample range
    MWCO
    Availability
    Price
  • 89369-470
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    6 - 8 K
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $100.49
    $100.49
    Pack of 10
       $100.49
    Supplier Number: IB48030
  • 89369-472
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    6 - 8 K
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $202.51
    $202.51
    Pack of 30
       $202.51
    Supplier Number: IB48035
  • 89369-478
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    6 - 8 K
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $311.77
    $311.77
    Pack of 50
       $311.77
    Supplier Number: IB48050
  • 89369-480
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    6 - 8 K
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $604.42
    $604.42
    Pack of 100
       $604.42
    Supplier Number: IB48060
  • 89369-474
    MINI FlexTube Kit, Tray
    MINI FlexTube Kit, Tray
    250 µl
    10 - 250 µl
    6 - 8 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $39.72
    $39.72
    Pack of 2
       $39.72
    Supplier Number: IB48040
  • 89369-476
    MINI FlexTube Kit, Float
    MINI FlexTube Kit, Float
    250 µl
    10 - 250 µl
    6 - 8 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $33.80
    $33.80
    Pack of 2
       $33.80
    Supplier Number: IB48041
  • 89369-486
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    12 - 14 K
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $61.79
    $61.79
    Pack of 10
       $61.79
    Supplier Number: IB48072
  • 89369-488
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    12 - 14 K
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $199.97
    $199.97
    Pack of 30
       $199.97
    Supplier Number: IB48073
  • 89369-490
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    12 - 14 K
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $310.28
    $310.28
    Pack of 50
       $310.28
    Supplier Number: IB48074
  • 89369-492
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    12 - 14 K
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $604.42
    $604.42
    Pack of 100
       $604.42
    Supplier Number: IB48075
  • 89369-482
    MINI FlexTube Kit, Tray
    MINI FlexTube Kit, Tray
    250 µl
    10 - 250 µl
    12 - 14 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $39.72
    $39.72
    Pack of 2
       $39.72
    Supplier Number: IB48070
  • 89369-484
    MINI FlexTube Kit, Float
    MINI FlexTube Kit, Float
    250 µl
    10 - 250 µl
    12 - 14 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $33.80
    $33.80
    Pack of 2
       $33.80
    Supplier Number: IB48071
  • 89369-498
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    25 K
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $142.66
    $142.66
    Pack of 10
       $142.66
    Supplier Number: IB48082
  • 89369-500
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    25 K
    Log in for availability
    Log in for availability
    Pack of 30
    Pack of 30 - $439.67
    $439.67
    Pack of 30
       $439.67
    Supplier Number: IB48083
  • 89369-502
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    25 K
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $711.75
    $711.75
    Pack of 50
       $711.75
    Supplier Number: IB48084
  • 89369-504
    MINI FlexTube Kit
    MINI FlexTube Kit
    250 µl
    10 - 250 µl
    25 K
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $1,407.31
    $1,407.31
    Pack of 100
       $1,407.31
    Supplier Number: IB48085
  • 89369-494
    MINI FlexTube Kit, Tray
    MINI FlexTube Kit, Tray
    250 µl
    10 - 250 µl
    25 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $51.52
    $51.52
    Pack of 2
       $51.52
    Supplier Number: IB48080
  • 89369-496
    MINI FlexTube Kit, Float
    MINI FlexTube Kit, Float
    250 µl
    10 - 250 µl
    25 K
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $44.13
    $44.13
    Pack of 2
       $44.13
    Supplier Number: IB48081
  • 89369-444
    MINI FlexTube Support Tray - 4 Place
    MINI FlexTube Support Tray - 4 Place
    Log in for availability
    Log in for availability
    Each
    Each - $18.76
    $18.76
    Each
       $18.76
    Supplier Number: IB48001
  • 89369-450
    MINI FlexTube Float - 9 Place
    MINI FlexTube Float - 9 Place
    Log in for availability
    Log in for availability
    Each
    Each - $9.01
    $9.01
    Each
       $9.01
    Supplier Number: IB48004