SMITH & WESSON® ELITE™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Lightweight 'retro-style' design provides the latest fashion statement.
- Wide temple design ensures structural integrity and stability
- Anti-scratch dual lenses with an 8-base curve provide wrap-around protection
- High performance polycarbonate lens provides 99.9% UV protection
Weight: 59 g (0.13 lbs.).
: Meets ANSI Z87.1+ standards.
: Supplied with micro-fiber carry pouch that doubles as a cleaning cloth.
Frequently Bought Together