ContactUsLinkComponent
KleenGuard™ Monogoggle™ 202 Safety Goggles, Kimberly-Clark Professional
KleenGuard™ Monogoggle™ 202 Safety Goggles, Kimberly-Clark Professional
Catalog # 33001-236
Supplier:  Avantor
undefined
KleenGuard™ Monogoggle™ 202 Safety Goggles, Kimberly-Clark Professional
Catalog # 33001-236
Supplier:  Avantor
Specifications
  • Frame color:
    Translucent green
  • Environmentally Preferable:
  • Cat. No.:
    33001-236
  • Lens coating:
    Anti-fog
  • Lens tint:
    Clear
  • Supplier no.:
    16667
  • Lens material:
    PC
Specifications
About this item
Offer workers protection from dust with the KleenGuard™ Monogoggle 202 safety goggles.
  • Unventilated design for protection from fine dust particles (D5 rating for fine dust protection and a D4 rating for dust protection)
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
  • Two-piece goggle frame with soft cushioned nose piece fits over most prescription eyeglasses
  • Anti-fog coating is ideal for temperature variances and humid environments
The unventilated design provides protection from fine dust particles. In fact, these safety goggles have a D5 rating for fine dust protection and a D4 rating for dust protection. The headband buckles are located toward the rear of the frame to limit interference with peripheral vision, and the goggles are made to fit over most prescription eyewear. Goggles are clear (with a green frame), making them ideal for indoor use and maximum light transmission, and they feature an anti-fog coating for humid environments.
Certifications: Meet the ANSI Z87.1+ standards for impact, with a D5 rating for fine dust protection and a D4 rating for dust protection.