Whizard® Stainless Steel Mesh Glove with 2" Cuff, Wells Lamont
Whizard® stainless steel gloves with 2" cuff are ergonomically designed to be worn on both hands.
- Ergonomically designed
- Patented self-adjusting tensioning system
- Micro plasma welded rings
- No gaps to trap soil
- Ambidextrous
- Easy to clean
Whizard® stainless steel gloves provide superior cut and puncture protection. They feature a patented self-adjusting tensioning system on wrist allows for an easy fit with no straps or buckles to fasten. All Whizard® mesh gloves come with a metal serial number tag, which helps with reissuance and sizing information. Micro plasma welded rings have no gaps to trap soil that can feed bacterial growth. These gloves are made with 4 mm ring using 0.5 mm wire. Gloves are ambidextrous; just turn inside out for the other hand. These gloves are easy to clean.
: These gloves are sold by the piece. Repair service is available to extend the life of the glove. For authorized repair center call 800-247-3295.
- Catalog No:
- 80091-072
- 80091-070
- 80091-066
- 80091-064
- 80091-068
- 80091-062
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable/Reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Cuff Length:
- 2"
- 2"
- 2"
- 2"
- 2"
- 2"
Frequently Bought Together