Phosphatase Inhibitor Cocktail I, Thermo Scientific
AAJ63907-AA
:
- Size:5 mL
- MDL No.:MFCD01635520
- Storage Temperature:-30°C to -10°C
- Physical Form:Liquid
- Appearance:Colorless
- Cat. No.:AAJ63907-AA
- Supplier no.:J63907-AA
Liquid
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD01635520