RIPA buffer with Triton® X-100 (5X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Specifications
- Storage Temperature:2°C to 8°C
- Physical Form:Liquid
Specifications
Related Information
Frequently Bought Together