TE buffer, pH 8.1, RNAse free, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Physical Form: Liquid
MDL No.: MFCD00236359
Specifications
- MDL No.:MFCD00236359
- Physical Form:Liquid
Specifications
Related Information
Frequently Bought Together