Bovine Desoxyribonuclease I (DNase I) (from Pancreas)
MW = 31 kDa.
- CAS No.:9003-98-9
- Source:Pancreas
- Species:Bovine
- Storage Conditions:2...8 °C
- Form:White to off-white lyophilized powder
- MDL No.:MFCD00130918
- Merck Reference:14,2906
- Enzyme Name:Desoxyribonuclease I (DNase I)
- Molecular Weight:31 kDa
Frequently Bought Together