About this item
A plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts.
- EINECS: 234-258-2
- GHS symbol and siganl word: GHS08 and warning
- Merck index number: 14,2495
CAS No.: 11028-71-0 Storage Temperature: 2°C to 8°C Physical Form: Lyophilized powder Appearance: White Solubility: Soluble in 0.9% saline or PBS MDL No.: MFCD00071069
Applications: A plant lectin that is a T-cell mitogen, and induces apoptosis in human fibroblasts
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:11028-71-0
- Storage Conditions:Ambient
- Form:Lyophilized powder
- MDL No.:MFCD00071069
- Protein/Peptide Name:Concanavalin A
Specifications
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Organic Building Blocks and Intermediates