NP-40 lysis buffer, low salt, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together