TRIS, 1.0 M Buffer Solutions, 0.2 µm Filtered, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 77-86-1
Odor: Odorless
MDL No.: MFCD00004679
Specifications
- CAS No.:77-86-1
- MDL No.:MFCD00004679
- Physical Form:Liquid
- Odor/Scent:Odorless
Specifications
Related Information
Frequently Bought Together