Potassium Phosphate, 0.2 M Buffer Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 7778-77-0
MDL No.: MFCD00011401
Specifications
- CAS No.:7778-77-0
- MDL No.:MFCD00011401
- Physical Form:Liquid
Specifications
Related Information
Frequently Bought Together