Wash buffer, 1.0% Triton® X-100, pH 8.0, Thermo Scientific
: Avantor
Liquid
Physical Form: Liquid
Appearance: Colorless
Applications: For GST-fusion protein purification
- Physical Form:Liquid
- Appearance:Colorless
Frequently Bought Together
Liquid
Physical Form: Liquid
Appearance: Colorless
Applications: For GST-fusion protein purification