Laemmli SDS sample buffer, reducing (4X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together