
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 0.5 M in water 95%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:51805-45-9
:
Formula:C₉H₁₆ClO₆P
MDL Number:MFCD00145469
:286.65 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Organic Building Blocks and Intermediates
Application Notes
Frequently Bought Together