Order Entry
United States
ContactUsLinkComponent
Shallow Four Slot Drawer Organizer, Polystyrene, TrippNT
Shallow Four Slot Drawer Organizer, Polystyrene, TrippNT
Catalog # 10012-712
Supplier:  TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Shallow Four Slot Drawer Organizer, Polystyrene, TrippNT
Catalog # 10012-712
Supplier:  TrippNT
Supplier Number:  50040
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Shallow Four-Slot Drawer Organizer
  • Cat. No.:
    10012-712
  • L×W×H:
    330x230x50 mm (13x9x2")
  • Weight:
    1 kg (2.2 lbs.)
  • Supplier no.:
    50040

Specifications

About this item

White polystyrene shallow 4 slot drawer organizer sorts, stores, protects and transports supplies

This drawer organizer is made from white, durable high impact styrene. Its rounded inside corners are easy to clean and allows for fast transportation of supplies. Designed specifically for shallow drawers or other low profile settings. The four equally sized compartments are great for holding syringes, needles, test tubes and other small supplies