ContactUsLinkComponent
Concentric Tube Column, Ace Glass Incorporated
Concentric Tube Column, Ace Glass Incorporated
Catalog # 89074-404
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Concentric Tube Column, Ace Glass Incorporated
Catalog # 89074-404
Supplier:  Ace Glass
Supplier Number:  9333-10
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Concentric Tube Column
  • Environmentally Preferable:
  • Cat. No.:
    89074-404
  • Supplier no.:
    9333-10
Specifications
About this item
This high efficiency, low holdup, low pressure drop fractionating column is vacuum jacketed with an observation stripe.
Suitable for samples as small as 2 ml. The center tube is removable for cleaning. Double expansion spiral eliminates choking. Joints are [ST] 14/20. Approximately 40 theoretical plates at 80 ml/hr. boil-up. Operating pressure drop less than 1 mm Hg. Column holdup (approx.): Operating — with a 89074-446 head, 0.6 ml at 80 ml/hr. Static — with a 89074-446 head, 0.25 ml Xylene. Static: Column alone, 0.1 ml Xylene. Fabricated from borosilicate glass.