Order Entry
United States
ContactUsLinkComponent
Dibromofluorescein (contains Mono-, Tri- and Tetra-)
Dibromofluorescein (contains Mono-, Tri- and Tetra-)
Catalog # TCD1120-005G
Supplier:  TCI America
CAS Number:  
Dibromofluorescein (contains Mono-, Tri- and Tetra-)
Catalog # TCD1120-005G
Supplier:  TCI America
Supplier Number:  D1120-5G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    5 g
  • Cat. No.:
    TCD1120-005G
  • Supplier no.:
    D1120-5G

Specifications

About this item

CAS Number:596-03-2
 :
Formula:C₂₀H₁₀Br₂O₅
MDL Number:MFCD00005042
 :490.10 g/mol